2-Bromo-4-fluorophenylacetylene

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-ethynyl-4-fluorobenzene

PC49260-1G PC49260-250MG PC49260-5G
APOSPC49260-1GEA 248 EUR
APOSPC49260-1G APOSPC49260-250MG APOSPC49260-5G
2-Bromo-4-fluorophenylacetylene
2-Bromo-4-fluorophenylacetylene
Formel: C₈H₄BrF
Molecular Weight: 199,02 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08703532
CAS-Nummer: 1057670-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR