5-Chloro-2,4-difluorobenzonitrile 97%

Lieferant: Apollo Scientific
Achtung

PC49273-1G PC49273-5G PC49273-10G
APOSPC49273-1GEA 24.8 EUR
APOSPC49273-1G APOSPC49273-5G APOSPC49273-10G
5-Chloro-2,4-difluorobenzonitrile 97%
5-Chloro-2,4-difluorobenzonitrile
Formel: C₇H₂ClF₂N
Molecular Weight: 173.547 g/mol
Schmelzpunkt: 48…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659465
CAS-Nummer: 146780-26-9
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR