6-Fluorobenzo[b]thiophene-2-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Fluoro-benzo[b]thiophene-2-carboxylic acid, 6-Fluoro-1-benzothiophene-2-carboxylic acid, 2-Carboxy-6-fluorobenzo[b]thiophene

PC49280-1G PC49280-250MG
APOSPC49280-1GEA 284 EUR
APOSPC49280-1G APOSPC49280-250MG
6-Fluorobenzo[b]thiophene-2-carboxylic acid 95%
6-Fluorobenzo[b]thiophene-2-carboxylic acid
Formel: C₉H₅FO₂S
Molecular Weight: 196,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06655861
CAS-Nummer: 142329-23-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR