1,1,1,2,2,3,3,4,4-Nonafluorononane 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Perfluorobut-1-yl)pentane

PC49286-1G PC49286-25G PC49286-5G
APOSPC49286-1GEA 67.2 EUR
APOSPC49286-1G APOSPC49286-25G APOSPC49286-5G
1,1,1,2,2,3,3,4,4-Nonafluorononane 97%
1,1,1,2,2,3,3,4,4-Nonafluorononane
Formel: C₉H₁₁F₉
Molecular Weight: 290.169 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22123953
CAS-Nummer: 1190430-21-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR