6-Bromo-4-fluoro-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

PC49288-1G PC49288-250MG PC49288-5G
APOSPC49288-1GEA 348 EUR
APOSPC49288-1G APOSPC49288-250MG APOSPC49288-5G
6-Bromo-4-fluoro-1H-benzimidazole
6-Bromo-4-fluoro-1H-benzimidazole
Formel: C₇H₄BrFN₂
Molecular Weight: 215,022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22558088
CAS-Nummer: 1197944-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR