2-Bromo-4-(trifluoromethoxy)benzeneboronic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Bromo-4-(trifluoromethoxy)phenyl)boronic acid
PC49295-1G
PC49295-250MG
PC49295-5G
APOSPC49295-1GEA
143
EUR
APOSPC49295-1G
APOSPC49295-250MG
APOSPC49295-5G
2-Bromo-4-(trifluoromethoxy)benzeneboronic acid
2-Bromo-4-(trifluoromethoxy)benzeneboronic acid
Formel:
C₇H₅BBrF₃O₃ Molecular Weight: 284,823 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD19237163 CAS-Nummer: 959997-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...