6-Bromo-4-(trifluoromethoxy)-1H-benzimidazole
Lieferant: Apollo Scientific
|
Achtung
|
PC49299-1G
PC49299-250MG
APOSPC49299-1GEA
441
EUR
APOSPC49299-1G
APOSPC49299-250MG
6-Bromo-4-(trifluoromethoxy)-1H-benzimidazole
6-Bromo-4-(trifluoromethoxy)-1H-benzimidazole
Formel:
C₈H₄BrF₃N₂O Molecular Weight: 281,029 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22628766 CAS-Nummer: 1417341-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...