2-Amino-5-(trifluoromethoxy)benzonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Trifluoromethoxy)anthranilonitrile, 2-Cyano-4-(trifluoromethoxy)aniline

PC49304-1G PC49304-5G
APOSPC49304-1GEA 187 EUR
APOSPC49304-1G APOSPC49304-5G
2-Amino-5-(trifluoromethoxy)benzonitrile
2-Amino-5-(trifluoromethoxy)benzonitrile
Formel: C₈H₅F₃N₂O
Molecular Weight: 202.133 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06408813
CAS-Nummer: 549488-77-9
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR