4-(Hydroxymethyl)-2-(trifluoromethyl)pyridine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-(Trifluoromethyl)pyridin-4-yl)methanol, [2-(Trifluoromethyl)pyridin-4-yl]methanol, 2-(Trifluoromethyl)isonicotinyl alcohol

PC49314-1G PC49314-250MG PC49314-500MG PC49314-5G
APOSPC49314-1GEA 311 EUR
APOSPC49314-1G APOSPC49314-250MG APOSPC49314-500MG APOSPC49314-5G
4-(Hydroxymethyl)-2-(trifluoromethyl)pyridine 95%
4-(Hydroxymethyl)-2-(trifluoromethyl)pyridine
Formel: C₇H₆F₃NO
Molecular Weight: 177,124 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100737
CAS-Nummer: 131747-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR