2-Amino-5-bromo-4-fluorobenzonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-4-fluoroanthranilonitrile, 4-Bromo-2-cyano-5-fluoroaniline
PC49323-1G
PC49323-250MG
PC49323-5G
APOSPC49323-1GEA
52.2
EUR
APOSPC49323-1G
APOSPC49323-250MG
APOSPC49323-5G
2-Amino-5-bromo-4-fluorobenzonitrile 97%
2-Amino-5-bromo-4-fluorobenzonitrile
Formel:
C₇H₄BrFN₂ Molecular Weight: 215,022 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD23098943 CAS-Nummer: 1334331-01-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...