3-Bromo-2-fluorostyrene 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-3-ethenyl-2-fluorobenzene, 1-Bromo-2-fluoro-3-vinylbenzene

PC49324-1G PC49324-250MG PC49324-5G
APOSPC49324-1GEA 325 EUR
APOSPC49324-1G APOSPC49324-250MG APOSPC49324-5G
3-Bromo-2-fluorostyrene 95%
3-Bromo-2-fluorostyrene
Formel: C₈H₆BrF
Molecular Weight: 201,036 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD23098944
CAS-Nummer: 871223-88-0
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR