2-Amino-6-bromo-4-[(trifluoromethyl)thio]phenol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-6-bromo-4-[(trifluoromethyl)sulphanyl]phenol, 3-Bromo-2-hydroxy-5-[(trifluoromethyl)thio]aniline

PC49347-1G PC49347-250MG
APOSPC49347-1GEA 859 EUR
APOSPC49347-1G APOSPC49347-250MG
2-Amino-6-bromo-4-[(trifluoromethyl)thio]phenol 97%
2-Amino-6-bromo-4-[(trifluoromethyl)thio]phenol
Formel: C₇H₅BrF₃NOS
Molecular Weight: 288.085 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD24369703
CAS-Nummer: 1432075-81-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR