2-Methyl-4-(trifluoromethoxy)benzylamine

Lieferant: Apollo Scientific
Gefahr

Synonyme: [2-Methyl-4-(trifluoromethoxy)phenyl]methylamine, 2-(Aminomethyl)-5-(trifluoromethoxy)toluene

PC49352-1G PC49352-250MG
APOSPC49352-1GEA 174 EUR
APOSPC49352-1G APOSPC49352-250MG
2-Methyl-4-(trifluoromethoxy)benzylamine
2-Methyl-4-(trifluoromethoxy)benzylamine
Formel: C₉H₁₀F₃NO
Molecular Weight: 205,177 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06213547
CAS-Nummer: 771572-39-5
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR