2,2,2,2',6'-Pentafluoroacetophenone 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2,6-Difluorophenyl)-2,2,2-trifluoroethanone, 1-(2,6-Difluorophenyl)-2,2,2-trifluoroethan-1-one

PC49356-1G PC49356-5G
APOSPC49356-1GEA 140 EUR
APOSPC49356-1G APOSPC49356-5G
2,2,2,2',6'-Pentafluoroacetophenone 95%
2,2,2,2',6'-Pentafluoroacetophenone
Formel: C₈H₃F₅O
Molecular Weight: 210.101 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08447307
CAS-Nummer: 1092712-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR