2-Bromo-4-[(trifluoromethyl)thio]aniline 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-4-[(trifluoromethyl)sulphanyl]aniline, 4-Amino-3-bromophenyl trifluoromethyl sulphide

PC49362-1G PC49362-250MG
APOSPC49362-1GEA 209 EUR
APOSPC49362-1G APOSPC49362-250MG
2-Bromo-4-[(trifluoromethyl)thio]aniline 97%
2-Bromo-4-[(trifluoromethyl)thio]aniline
Formel: C₇H₅BrF₃NS
Molecular Weight: 272,086 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD24850034
CAS-Nummer: 940077-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR