2,3-Diamino-6-fluorobenzotrifluoride 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluoro-3-(trifluoromethyl)benzene-1,2-diamine, 4-Fluoro-3-(trifluoromethyl)phenylene-1,2-diamine
PC49367-1G
PC49367-250MG
PC49367-5G
APOSPC49367-1GEA
46.4
EUR
APOSPC49367-1G
APOSPC49367-250MG
APOSPC49367-5G
2,3-Diamino-6-fluorobenzotrifluoride 95%
2,3-Diamino-6-fluorobenzotrifluoride
Formel:
C₇H₆F₄N₂ Molecular Weight: 194,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD24850041 CAS-Nummer: 1440535-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...