5-Fluoro-2-iodobenzyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: α-Bromo-5-fluoro-2-iodotoluene, 2-(Bromomethyl)-4-fluoro-1-iodobenzene

PC49376-1G PC49376-250MG
APOSPC49376-1GEA 963 EUR
APOSPC49376-1G APOSPC49376-250MG
5-Fluoro-2-iodobenzyl bromide 97%
5-Fluoro-2-iodobenzyl bromide
Formel: C₇H₅BrFI
Molecular Weight: 314,921 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18396961
CAS-Nummer: 1261852-14-5
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR