2-Iodo-6-nitro-4-(trifluoromethoxy)phenol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Hydroxy-3-iodo-5-(trifluoromethoxy)nitrobenzene

PC49383-1G PC49383-250MG
APOSPC49383-1GEA 604 EUR
APOSPC49383-1G APOSPC49383-250MG
2-Iodo-6-nitro-4-(trifluoromethoxy)phenol
2-Iodo-6-nitro-4-(trifluoromethoxy)phenol
Formel: C₇H₃F₃INO₄
Molecular Weight: 349,003 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27665000
CAS-Nummer: 1820607-53-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR