2-Amino-5-bromo-3-(trifluoromethyl)benzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC49385-1G
PC49385-250MG
PC49385-5G
APOSPC49385-1GEA
238
EUR
APOSPC49385-1G
APOSPC49385-250MG
APOSPC49385-5G
2-Amino-5-bromo-3-(trifluoromethyl)benzoic acid
2-Amino-5-bromo-3-(trifluoromethyl)benzoic acid
Formel:
C₈H₅BrF₃NO₂ Molecular Weight: 284,03 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19441382 CAS-Nummer: 50419-85-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...