2-Bromo-3-(trifluoromethyl)benzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-1-(bromomethyl)-3-(trifluoromethyl)benzene

PC49388-1G PC49388-5G PC49388-25G
APOSPC49388-1GEA 134 EUR
APOSPC49388-1G APOSPC49388-5G APOSPC49388-25G
2-Bromo-3-(trifluoromethyl)benzyl bromide 95%
2-Bromo-3-(trifluoromethyl)benzyl bromide
Formel: C₈H₅Br₂F₃
Molecular Weight: 317,929 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185412
CAS-Nummer: 1214372-35-6
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR