2-Bromo-3-(trifluoromethyl)benzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC49389-1G
PC49389-250MG
PC49389-5G
APOSPC49389-1GEA
168
EUR
APOSPC49389-1G
APOSPC49389-250MG
APOSPC49389-5G
2-Bromo-3-(trifluoromethyl)benzaldehyde 95%
2-Bromo-3-(trifluoromethyl)benzaldehyde
Formel:
C₈H₄BrF₃O Molecular Weight: 253,016 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11520657 CAS-Nummer: 1114808-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...