8-Fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC49395-1G
PC49395-250MG
APOSPC49395-1GEA
313
EUR
APOSPC49395-1G
APOSPC49395-250MG
8-Fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
8-Fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
Formel:
C₈H₅FN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09994934 CAS-Nummer: 1020034-56-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...