6-Bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC49399-1G
PC49399-250MG
APOSPC49399-1GEA
192
EUR
APOSPC49399-1G
APOSPC49399-250MG
6-Bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
6-Bromo-8-fluoroimidazo[1,2-a]pyridine-2-carboxylic acid
Formel:
C₈H₄BrFN₂O₂ Molecular Weight: 259,032 g/mol Schmelzpunkt: 240…243 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD13193299 CAS-Nummer: 1260784-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...