Ethyl 3-bromo-7-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC49403-1G
PC49403-250MG
APOSPC49403-1GEA
208
EUR
APOSPC49403-1G
APOSPC49403-250MG
Ethyl 3-bromo-7-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 3-bromo-7-fluoroimidazo[1,2-a]pyridine-2-carboxylate
Formel:
C₁₀H₈BrFN₂O₂ Molecular Weight: 287,085 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31745721 CAS-Nummer: 2379918-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...