2-Cyclopropyl-1-fluoro-3-nitrobenzene

Lieferant: Apollo Scientific
Achtung

PC49409-1G PC49409-250MG PC49409-5G
APOSPC49409-1GEA 30.6 EUR
APOSPC49409-1G APOSPC49409-250MG APOSPC49409-5G
2-Cyclopropyl-1-fluoro-3-nitrobenzene
2-Cyclopropyl-1-fluoro-3-nitrobenzene
Formel: C₉H₈FNO₂
Molecular Weight: 181,164 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD31793565
CAS-Nummer: 2366994-55-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR