5-Cyclopropyl-2-fluoroaniline
Lieferant: Apollo Scientific
|
Achtung
|
PC49417-1G
PC49417-250MG
PC49417-5G
APOSPC49417-1GEA
0
EUR
APOSPC49417-1G
APOSPC49417-250MG
APOSPC49417-5G
5-Cyclopropyl-2-fluoroaniline
5-Cyclopropyl-2-fluoroaniline
Formel:
C₉H₁₀FN Lagertemperatur: Gefrierschrank |
MDL:
MFCD24645335 CAS-Nummer: 1374298-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...