3-Cyclopropyl-5-fluoroaniline
Lieferant: Apollo Scientific
|
Achtung
|
PC49421-1G
PC49421-250MG
PC49421-5G
APOSPC49421-1GEA
104
EUR
APOSPC49421-1G
APOSPC49421-250MG
APOSPC49421-5G
3-Cyclopropyl-5-fluoroaniline
3-Cyclopropyl-5-fluoroaniline
Formel:
C₉H₁₀FN Molecular Weight: 151.181 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD30210561 CAS-Nummer: 1448348-37-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...