5-(4-(Trifluoromethoxy)phenyl)thiophene-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC49428-1G
PC49428-250MG
PC49428-5G
APOSPC49428-1GEA
209
EUR
APOSPC49428-1G
APOSPC49428-250MG
APOSPC49428-5G
5-(4-(Trifluoromethoxy)phenyl)thiophene-2-carboxylic acid
5-(4-(Trifluoromethoxy)phenyl)thiophene-2-carboxylic acid
Formel:
C₁₂H₇F₃O₃S Molecular Weight: 288,242 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12739572 CAS-Nummer: 1039859-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...