4-(Difluoromethyl)-2-(trifluoromethyl)benzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC49446-1G
PC49446-250MG
PC49446-5G
APOSPC49446-1GEA
26.1
EUR
APOSPC49446-1G
APOSPC49446-250MG
APOSPC49446-5G
4-(Difluoromethyl)-2-(trifluoromethyl)benzoic acid 95%
4-(Difluoromethyl)-2-(trifluoromethyl)benzoic acid
Formel:
C₉H₅F₅O₂ Molecular Weight: 240,127 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31759994 CAS-Nummer: 2248272-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...