1-(6-Bromo-4-chloro-2,3-difluoro-phenyl)ethanone 95%

Lieferant: Apollo Scientific

PC49472-1G PC49472-250MG
APOSPC49472-1GEA 609 EUR
APOSPC49472-1G APOSPC49472-250MG
1-(6-Bromo-4-chloro-2,3-difluoro-phenyl)ethanone 95%
1-(6-Bromo-4-chloro-2,3-difluoro-phenyl)ethanone
Formel: C₈H₄BrClF₂O
Molecular Weight: 269,471 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2416368-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR