5-Bromo-2,3,4-trifluorobenzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-Bromo-5-(bromomethyl)-2,3,4-trifluorobenzene

PC49489-1G PC49489-250MG PC49489-5G
APOSPC49489-1GEA 58 EUR
APOSPC49489-1G APOSPC49489-250MG APOSPC49489-5G
5-Bromo-2,3,4-trifluorobenzyl bromide 95%
5-Bromo-2,3,4-trifluorobenzyl bromide
Formel: C₇H₃Br₂F₃
Molecular Weight: 303.902 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12756772
CAS-Nummer: 1260832-85-6
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR