2-Hydroxy-6-(trifluoromethyl)benzaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-(Trifluoromethyl)salicylaldehyde, 2-Formyl-3-hydroxybenzotrifluoride, 2-Formyl-3-(trifluoromethyl)phenol
PC49535-1G
PC49535-250MG
PC49535-5G
APOSPC49535-1GEA
77
EUR
APOSPC49535-1G
APOSPC49535-250MG
APOSPC49535-5G
2-Hydroxy-6-(trifluoromethyl)benzaldehyde 95%
2-Hydroxy-6-(trifluoromethyl)benzaldehyde
Formel:
C₈H₅F₃O₂ Molecular Weight: 190,119 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD16999701 CAS-Nummer: 58914-35-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...