1-(2-Fluorophenyl)cyclopropan-1-amine hydrochloride 96%
Lieferant: Apollo Scientific
Synonyme:
1-Amino-1-(2-fluorophenyl)cyclopropane hydrochloride, 1-(1-Aminocycloprop-1-yl)-2-fluorobenzene hydrochloride
PC49539-10G
PC49539-1G
PC49539-5G
APOSPC49539-10GEA
1520
EUR
APOSPC49539-10G
APOSPC49539-1G
APOSPC49539-5G
1-(2-Fluorophenyl)cyclopropan-1-amine hydrochloride 96%
1-(2-Fluorophenyl)cyclopropan-1-amine hydrochloride
Formel:
C₉H₁₀FN·ClH Molecular Weight: 187,642 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09028126 CAS-Nummer: 1215107-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...