5-Bromo-6-fluoro-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-6-fluoro-1H-benzo[d]imidazole

PC49540-1G PC49540-250MG
APOSPC49540-1GEA 102 EUR
APOSPC49540-1G APOSPC49540-250MG
5-Bromo-6-fluoro-1H-benzimidazole
5-Bromo-6-fluoro-1H-benzimidazole
Formel: C₇H₄BrFN₂
Molecular Weight: 215.022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11846341
CAS-Nummer: 1008360-84-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR