2-Fluoroallyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Fluoroprop-2-en-1-ol, 2-Fluoro-3-hydroxyprop-1-ene

PC49591-5G
APOSPC49591-5GEA 0 EUR
APOSPC49591-5G
2-Fluoroallyl alcohol
2-Fluoroallyl alcohol
Formel: C₃H₅FO
Molecular Weight: 76,07 g/mol
Siedepunkt: 35…37 °C (1 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD01696257
CAS-Nummer: 5675-31-0
UN: 1992
ADR: 3+6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR