5-Bromo-6-fluoro-1,3-benzoxazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-6-fluorobenzo[d]oxazole

PC49597-1G PC49597-250MG
APOSPC49597-1GEA 580 EUR
APOSPC49597-1G APOSPC49597-250MG
5-Bromo-6-fluoro-1,3-benzoxazole
5-Bromo-6-fluoro-1,3-benzoxazole
Formel: C₇H₃BrFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD24850044
CAS-Nummer: 1440535-10-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR