4-Fluoro-3-iodoaniline

Lieferant: Apollo Scientific
Achtung

PC49605-1G PC49605-250MG PC49605-5G
APOSPC49605-1GEA 122 EUR
APOSPC49605-1G APOSPC49605-250MG APOSPC49605-5G
4-Fluoro-3-iodoaniline
4-Fluoro-3-iodoaniline
Formel: C₆H₅FIN
Molecular Weight: 237,013 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18396914
CAS-Nummer: 647025-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR