2-Hydroxy-3-(trifluoromethyl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Trifluoromethyl)salicylic acid, 3-Carboxy-2-hydroxybenzotrifluoride

PC49616-1G PC49616-5G PC49616-25G
APOSPC49616-1GEA 112 EUR
APOSPC49616-1G APOSPC49616-5G APOSPC49616-25G
2-Hydroxy-3-(trifluoromethyl)benzoic acid 95%
2-Hydroxy-3-(trifluoromethyl)benzoic acid
Formel: C₈H₅F₃O₃
Schmelzpunkt: 161…163 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06203614
CAS-Nummer: 251300-32-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR