3-Fluoro-2-methoxyphenacyl bromide 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromo-3'-fluoro-2'-methoxyacetophenone, 2-(Bromoacetyl)-6-fluoroanisole, 2-Bromo-1-(3-fluoro-2-methoxyphenyl)ethan-1-one
PC49619-1G
PC49619-250MG
APOSPC49619-1GEA
261
EUR
APOSPC49619-1G
APOSPC49619-250MG
3-Fluoro-2-methoxyphenacyl bromide 97%
3-Fluoro-2-methoxyphenacyl bromide
Formel:
C₉H₈BrFO₂ Molecular Weight: 247,061 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD23711719 CAS-Nummer: 1427403-76-6 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...