5-Amino-2-bromo-4-fluorophenol 95%

Lieferant: Apollo Scientific
Achtung

PC49626-1G PC49626-250MG PC49626-5G
APOSPC49626-1GEA 574 EUR
APOSPC49626-1G APOSPC49626-250MG APOSPC49626-5G
5-Amino-2-bromo-4-fluorophenol 95%
5-Amino-2-bromo-4-fluorophenol
Formel: C₆H₅BrFNO
Lagertemperatur: Raumtemperatur
MDL: MFCD18762025
CAS-Nummer: 84478-73-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR