3-Chloro-5-iodobenzotrifluoride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-5-iodo-α,α,α-trifluorotoluene, 1-Chloro-3-iodo-5-(trifluoromethyl)benzene

PC49629-25G
APOSPC49629-25GEA 119 EUR
APOSPC49629-25G
3-Chloro-5-iodobenzotrifluoride
3-Chloro-5-iodobenzotrifluoride
Formel: C₇H₃ClF₃I
Molecular Weight: 306,451 g/mol
Siedepunkt: 98…112 °C (15 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD19707625
CAS-Nummer: 1189352-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR