4-Bromo-3,5-difluorobenzenesulphonyl chloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Bromo-3,5-difluorobenzene-1-sulphonyl chloride
PC4965-1G
PC4965-5G
PC4965-25G
APOSPC4965-1GEA
21.9
EUR
APOSPC4965-1G
APOSPC4965-5G
APOSPC4965-25G
4-Bromo-3,5-difluorobenzenesulphonyl chloride 97%
4-Bromo-3,5-difluorobenzenesulphonyl chloride
Formel:
C₆H₂BrClF₂O₂S Molecular Weight: 291,498 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788548 CAS-Nummer: 518057-63-1 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...