2-Bromo-5-fluoro-4-iodotoluene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-4-fluoro-5-iodo-2-methylbenzene

PC49650-1G PC49650-250MG
APOSPC49650-1GEA 279 EUR
APOSPC49650-1G APOSPC49650-250MG
2-Bromo-5-fluoro-4-iodotoluene
2-Bromo-5-fluoro-4-iodotoluene
Formel: C₇H₅BrFI
Molecular Weight: 314,921 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28040490
CAS-Nummer: 1805937-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR