2,6-Difluoro-4-(trifluoromethyl)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: [2,6-Difluoro-4-(trifluoromethyl)phenyl]methanol

PC49664-1G PC49664-250MG PC49664-5G
APOSPC49664-1GEA 720 EUR
APOSPC49664-1G APOSPC49664-250MG APOSPC49664-5G
2,6-Difluoro-4-(trifluoromethyl)benzyl alcohol
2,6-Difluoro-4-(trifluoromethyl)benzyl alcohol
Formel: C₈H₅F₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD28012313
CAS-Nummer: 1803570-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR