3-(Difluoromethoxy)toluene 98%

Lieferant: Apollo Scientific

Synonyme: 1-(Difluoromethoxy)-3-methylbenzene

PC49718-1G PC49718-250MG
APOSPC49718-1GEA 464 EUR
APOSPC49718-1G APOSPC49718-250MG
3-(Difluoromethoxy)toluene 98%
3-(Difluoromethoxy)toluene
Formel: C₈H₈F₂O
Molecular Weight: 158,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185566
CAS-Nummer: 4837-15-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR