2,2,2-Trifluoroethyl-2-fluoroacrylate
Lieferant: Apollo Scientific
|
Achtung
|
PC4975-1G
PC4975-250MG
APOSPC4975-1GEA
0
EUR
APOSPC4975-1G
APOSPC4975-250MG
2,2,2-Trifluoroethyl-2-fluoroacrylate
2,2,2-Trifluoroethyl-2-fluoroacrylate
Formel:
C₅H₄F₄O₂ Siedepunkt: 28 °C (30 mm Hg) Lagertemperatur: Kühlschrank |
MDL:
MFCD04039290 CAS-Nummer: 74359-10-7 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...