2-Bromo-4-chloro-5-fluorobenzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC49811-1G
PC49811-250MG
APOSPC49811-1GEA
638
EUR
APOSPC49811-1G
APOSPC49811-250MG
2-Bromo-4-chloro-5-fluorobenzoic acid 95%
2-Bromo-4-chloro-5-fluorobenzoic acid
Formel:
C₇H₃BrClFO₂ Molecular Weight: 253.453 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28733901 CAS-Nummer: 157165-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...