2-Bromo-5-fluoro-4-(trifluoromethyl)aniline

Lieferant: Apollo Scientific
Achtung

PC49825-1G PC49825-5G
APOSPC49825-1GEA 429 EUR
APOSPC49825-1G APOSPC49825-5G
2-Bromo-5-fluoro-4-(trifluoromethyl)aniline
2-Bromo-5-fluoro-4-(trifluoromethyl)aniline
Formel: C₇H₄BrF₄N
Molecular Weight: 258,011 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22200127
CAS-Nummer: 1239463-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR