2-Fluoro-6-nitrophenylacetonitrile

Lieferant: Apollo Scientific

PC499016-1G PC499016-250MG PC499016-5G
APOSPC499016-1GEA 288 EUR
APOSPC499016-1G APOSPC499016-250MG APOSPC499016-5G
2-Fluoro-6-nitrophenylacetonitrile
2-Fluoro-6-nitrophenylacetonitrile
Formel: C₈H₅FN₂O₂
Molecular Weight: 180.136 g/mol
Schmelzpunkt: 60…63.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11846360
CAS-Nummer: 105003-93-8
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR