4-Nitro-2-(trifluoromethyl)phenylacetonitrile
Lieferant: Apollo Scientific
PC499017-1G
PC499017-250MG
APOSPC499017-1GEA
393
EUR
APOSPC499017-1G
APOSPC499017-250MG
4-Nitro-2-(trifluoromethyl)phenylacetonitrile
4-Nitro-2-(trifluoromethyl)phenylacetonitrile
Formel:
C₉H₅F₃N₂O₂ Molecular Weight: 230,143 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185782 CAS-Nummer: 1067192-61-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...